Learn More
Medchemexpress LLC 4-benzhydryloxy-1-[3-(2H-tetrazol-5-yl)propyl]piperidine | 162641-16-9 | MFCD00936179 | 99.9% | 377.48 g·mol⁻¹ | C22H27N5O | 25 MG
SDP
Catalog No. 5000242457
Encompass_Preferred
HQL-79 is a potent, selective, and orally active inhibitor of human hematopoietic prostaglandin D synthase (H-PGDS) used as a research tool to study prostaglandin D2 synthesis, allergic responses, and inflammatory signaling. The compound is supplied at high purity with documented solubility and storage parameters to support reproducible experimental work.
  • Potent, selective inhibitor of hematopoietic prostaglandin D synthase (H-PGDS).
  • Orally active profile supports in vivo pharmacology studies.
  • High purity for reproducible biochemical and cellular assays.
  • Documented solubility in DMSO simplifies assay preparation.
  • Defined storage recommendations support long-term stability.
  • Available in small research pack sizes for screening and dose-ranging.
Catalog No. 50-002-42457 Supplier Medchemexpress LLC Supplier No. HY10825925MG
HQL-79 is a potent, selective, and orally active inhibitor of human hematopoietic prostaglandin D synthase (H-PGDS) used as a research tool to study prostaglandin D2 synthesis, allergic responses, and inflammatory signaling. The compound is supplied at high purity with documented solubility and storage parameters to support reproducible experimental work.
  • Potent, selective inhibitor of hematopoietic prostaglandin D synthase (H-PGDS).
  • Orally active profile supports in vivo pharmacology studies.
  • High purity for reproducible biochemical and cellular assays.
  • Documented solubility in DMSO simplifies assay preparation.
  • Defined storage recommendations support long-term stability.
  • Available in small research pack sizes for screening and dose-ranging.