Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-(Benzo[d]oxazol-2-yl)aniline | 20934-81-0 | MFCD00453046 | 99.9% | 210.24 | C13H10N2O | 25 G
SDP

Catalog No. 5000302850
Encompass_Preferred

4-(Benzo[d]oxazol-2-yl)aniline (CAS 20934-81-0) is a small-molecule research compound reported to have antitumor activity, including inhibitory effects against mammary carcinoma cell lines. The product is analytically characterized with HPLC purity reported at 99.93% and has defined storage recommendations to preserve stability.

  • High HPLC purity (99.93%) for reliable analytical work.
  • Reported antitumor activity against mammary carcinoma cell lines.
  • Molecular formula C13H10N2O; molecular weight 210.24.
  • Recommended storage: 4°C, protect from light; in solvent: -80°C (up to 6 months), -20°C (1 month).
  • Suitable as a research reagent for oncology and related studies.

Catalog No. 50-003-02850 Supplier Medchemexpress LLC Supplier No. 25GHYW029411
May include imposed supplier surcharges.
Add to Cart
Add to Cart

4-(Benzo[d]oxazol-2-yl)aniline (CAS 20934-81-0) is a small-molecule research compound reported to have antitumor activity, including inhibitory effects against mammary carcinoma cell lines. The product is analytically characterized with HPLC purity reported at 99.93% and has defined storage recommendations to preserve stability.

  • High HPLC purity (99.93%) for reliable analytical work.
  • Reported antitumor activity against mammary carcinoma cell lines.
  • Molecular formula C13H10N2O; molecular weight 210.24.
  • Recommended storage: 4°C, protect from light; in solvent: -80°C (up to 6 months), -20°C (1 month).
  • Suitable as a research reagent for oncology and related studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.