Learn More
Medchemexpress LLC 4-(benzo[d]thiazol-2-ylthio)-3-chloroaniline | 300809-71-6 | MFCD00227023 | 99.9% | C13H9ClN2S2 | 10MG
SDP
Catalog No. 5000259075
Encompass_Preferred
KRAS inhibitor-9 is a small-molecule research compound that blocks formation of GTP-bound KRAS and downstream KRAS signaling, inducing G2/M cell cycle arrest and apoptosis in responsive cells. It selectively reduces proliferation of KRAS-mutant non-small cell lung cancer cells and is supplied as a high-purity solid or as a DMSO solution for biochemical and cellular assays.
  • Potent KRAS inhibition with measurable binding affinity.
  • Selectively inhibits proliferation of KRAS-mutant non-small cell lung cancer cells.
  • Induces G2/M cell cycle arrest and apoptosis in responsive cell lines.
  • High purity and suitable for in vitro biochemical and cell-based assays.
  • Available in multiple pack sizes and solution form for experimental convenience.
Catalog No. 50-002-59075 Supplier Medchemexpress LLC Supplier No. HY13749710MG
Add to Cart
Add to Cart
KRAS inhibitor-9 is a small-molecule research compound that blocks formation of GTP-bound KRAS and downstream KRAS signaling, inducing G2/M cell cycle arrest and apoptosis in responsive cells. It selectively reduces proliferation of KRAS-mutant non-small cell lung cancer cells and is supplied as a high-purity solid or as a DMSO solution for biochemical and cellular assays.
  • Potent KRAS inhibition with measurable binding affinity.
  • Selectively inhibits proliferation of KRAS-mutant non-small cell lung cancer cells.
  • Induces G2/M cell cycle arrest and apoptosis in responsive cell lines.
  • High purity and suitable for in vitro biochemical and cell-based assays.
  • Available in multiple pack sizes and solution form for experimental convenience.