Learn More
Medchemexpress LLC 4-benzoylbenzoic acid | 611-95-0 | MFCD00002560 | 99.0% | 226.23 g/mol | C14H10O3 | 25 G
SDP
Catalog No. 5000209671
Encompass_Preferred
4-Benzoylbenzoic acid is a photooxidant used as an oxidant in organic synthesis and in photoactivation studies for antibacterial and antiviral compound development. It is supplied as a white to off-white solid at high purity and is offered in multiple laboratory-scale package sizes.
Catalog No. 50-002-09671 Supplier Medchemexpress LLC Supplier No. 25GHYW041343
4-Benzoylbenzoic acid is a photooxidant used as an oxidant in organic synthesis and in photoactivation studies for antibacterial and antiviral compound development. It is supplied as a white to off-white solid at high purity and is offered in multiple laboratory-scale package sizes.