Learn More
4-Benzoylbenzoic acid, 98%
Description
4-Benzoylbenzoic acid,is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field etc.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Benzoylbenzoic acid,is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field etc.
Solubility
It is soluble in water (partly).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.Incompatible with strong bases, strong oxidizing agents.
Specifications
Specifications
| Melting Point | 198°C to 200°C |
| Odor | Odorless |
| Quantity | 5 g |
| Beilstein | 1960224 |
| Formula Weight | 226.23 |
| Percent Purity | 99% |
| Chemical Name or Material | 4-Benzoylbenzoic acid |
Safety and Handling
| missing translation for 'healthHazard1' | Warning |
| missing translation for 'healthHazard2' | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| missing translation for 'healthHazard3' | GHS P Statement P280g-P305+P351+P338 Wear protective gloves. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. |
| RTECSNumber | DG3650000 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.