Learn More
Accela Chembio Inc 4-benzyl-5-oxomorpholine-3-carboxylic Acid | 5g | 106910-79-6 | MFCD11042914 | 97+% | Shelf Life: 1260 Days | Light Sensitive/+4
Supplier:  Accela Chembio Inc SY0051125G
Encompass_Preferred
4-benzyl-5-oxomorpholine-3-carboxylic Acid | 5g | 106910-79-6 | MFCD11042914 | 97+% | Shelf Life: 1260 Days | Light Sensitive/+4
Catalog No. 50-250-682