Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartate 98.0+%, TCI America™
SDP

Catalog No. B38875G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38875G 5 g
B388725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38875G Supplier TCI America Supplier No. B38875G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 86060-84-6
Molecular Formula C26H23NO6
Molecular Weight (g/mol) 445.47
MDL Number MFCD00065630
InChI Key OQGAELAJEGGNKG-UHFFFAOYNA-N
Synonym fmoc-asp obzl-oh, fmoc-l-aspartic acid 4-benzyl ester, l-fmoc-aspartic acid beta-benzyl ester, fmoc-l-asp obzl-oh, 2s-4-benzyloxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid, fmoc-l-aspartic acid beta-benzyl ester, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-benzyloxy-4-oxobutanoic acid, n-fmoc-l-aspartic acid beta-benzyl ester, fmoc-l-asp obn, fmoc-l-asp bzl-oh
PubChem CID 13966931
IUPAC Name 4-(benzyloxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-oxobutanoic acid
SMILES OC(=O)C(CC(=O)OCC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 127°C
Color White
Quantity 5 g
Formula Weight 445.47
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.