Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzylaniline, 98%

Catalog No. AAL1987303
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1987303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1987303 Supplier Thermo Scientific Chemicals Supplier No. L1987303
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1135-12-2 | C13H13N | 183.254 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1135-12-2
Molecular Formula C13H13N
Molecular Weight (g/mol) 183.254
MDL Number MFCD00047861
InChI Key WDTRNCFZFQIWLM-UHFFFAOYSA-N
Synonym 4-aminodiphenylmethane, benzenamine, 4-phenylmethyl, 4-benzyl-phenylamine, 4-benzylbenzenamine, 4-benzylphenylamine, 4-benzylaniline, hcl, 4-benzyl aniline, 4-benzyl-aniline, #, p-aminodiphenylmethan
PubChem CID 136914
IUPAC Name 4-benzylaniline
SMILES C1=CC=C(C=C1)CC2=CC=C(C=C2)N

Specifications

Melting Point 35°C to 38°C
Boiling Point 142°C to 144°C (1 mmHg)
Quantity 1 g
Beilstein 2208463
Formula Weight 183.25
Percent Purity 98%
Chemical Name or Material 4-Benzylaniline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.