Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzylbiphenyl 99.0+%, TCI America™
SDP

Catalog No. B151925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B151925G 25 g
B15195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B151925G Supplier TCI America Supplier No. B151925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 613-42-3
Molecular Formula C19H16
Molecular Weight (g/mol) 244.34
MDL Number MFCD00041831
InChI Key AGPLQTQFIZBOLI-UHFFFAOYSA-N
Synonym 4-benzylbiphenyl, 4-phenylmethyl-1,1'-biphenyl, 1,1'-biphenyl, 4-phenylmethyl, benzylbiphenyl, p-benzylbiphenyl, 4-phenyldiphenylmethane, 1-phenyl-4-benzylbenzene, 1, 4-phenylmethyl, acmc-1bh4u, 1-benzyl-4-phenyl-benzene
PubChem CID 69174
IUPAC Name 4-benzyl-1,1'-biphenyl
SMILES C(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 86°C
Color White
Boiling Point 286°C
Quantity 25 g
Formula Weight 244.34
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzylbiphenyl
EINECSNumber (4)-1501
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.