Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzyloxy-2-fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. B4638200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4638200MG 200 mg
B46381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4638200MG Supplier TCI America Supplier No. B4638200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 166744-78-1
Molecular Formula C13H12BFO3
Molecular Weight (g/mol) 246.044
MDL Number MFCD02093064
InChI Key PQWKGFALWCOVTC-UHFFFAOYSA-N
Synonym 4-benzyloxy-2-fluorophenylboronic acid, 4-benzyloxy-2-fluorobenzeneboronic acid, 4-benzyloxy-2-fluorophenyl boronic acid, boronic acid, 2-fluoro-4-phenylmethoxy phenyl, 2-fluoro-4-phenylmethoxyphenyl boronic acid, 4-benzyloxy-2-fluorophenylboronic acid, may contain varying amounts of anhydride, pubchem7760, akos brn-0077, acmc-209dv0
PubChem CID 2737786
IUPAC Name (2-fluoro-4-phenylmethoxyphenyl)boronic acid
SMILES B(C1=C(C=C(C=C1)OCC2=CC=CC=C2)F)(O)O

Specifications

Melting Point 155°C
Color White
Quantity 200 mg
Formula Weight 246.04
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzyloxy-2-fluorophenylboronic Acid (contains varying amounts of Anhydride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.