Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzyloxy-2-methoxybenzaldehyde, 98+%, Thermo Scientific™

Catalog No. AAL1551603
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1551603 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1551603 Supplier Thermo Scientific Chemicals Supplier No. L1551603

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 58026-14-5
Boiling Point ∼180°C (2mmHg)
Molecular Formula C15H14O3
Quantity 1g
MDL Number MFCD01075699
Beilstein 1880700
Synonym 4-benzyloxy-2-methoxybenzaldehyde, 2-methoxy-4-benzyloxybenzaldehyde, benzaldehyde, 2-methoxy-4-phenylmethoxy, 2-methoxy-4-phenylmethoxy benzaldehyde, pubchem3931, acmc-1awka, ksc496e3t, 5-benzyloxy-2-formylanisole, 4-benzyloxy-2-methoxy-benzaldehyde
InChI Key HZLNIOUBXRIFML-UHFFFAOYSA-N
SMILES COC1=C(C=CC(=C1)OCC2=CC=CC=C2)C=O
IUPAC Name 2-methoxy-4-phenylmethoxybenzaldehyde
Molecular Weight (g/mol) 242.274
PubChem CID 4293990
Formula Weight 242.27
Percent Purity ≥98%
Chemical Name or Material 4-Benzyloxy-2-methoxybenzaldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.