Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzyloxy-3-methoxyphenylacetic Acid 98.0+%, TCI America™
SDP

Catalog No. B48005G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48005G 5 g
B48001G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48005G Supplier TCI America Supplier No. B48005G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 29973-91-9
Molecular Formula C16H16O4
Molecular Weight (g/mol) 272.3
MDL Number MFCD00185562
InChI Key AYCIHUSEBJLTBF-UHFFFAOYSA-N
PubChem CID 297964
IUPAC Name 2-(3-methoxy-4-phenylmethoxyphenyl)acetic acid
SMILES COC1=C(C=CC(=C1)CC(=O)O)OCC2=CC=CC=C2

Specifications

Melting Point 118°C
Color White-Yellow
Quantity 5 g
Formula Weight 272.30
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzyloxy-3-methoxyphenylacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.