Learn More
Sigma Aldrich 4-(benzyloxy)benzene-1-sulfonyl chloride
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- (bromomethyl)cyclobutane (bromomethyl-cyclobutane) is an aryl fluorinated building block
- Its reaction with various hydroxychromen-4-one derivatives in the presence of anhydrous k2co3/dry acetone and bu4n+i- (ptc) has been investigated.(bromomethyl)cyclobutane may be used in the synthesis of racemic 3-cyclobutylalanine and 17-(cyclobutylmethyl)morphinan-3-ol (butorphan)
Specifications
Specifications
| Molecular Formula | C13H11ClO3S |
| Linear Formula | C13H11ClO3S |
| MDL Number | MFCD04035557 |
| Quantity | 5 mL |
| Molecular Weight (g/mol) | 282.74 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.