Learn More
Medchemexpress LLC 4-(Benzyloxy)butanal | 5470-84-8 | MFCD11553544 | 95.0% | 178.23 g/mol | C11H14O2 | 500 MG
SDP
Catalog No. 5000283715
Encompass_Preferred
4-(Benzyloxy)butanal is a benzyloxy-substituted aliphatic aldehyde used as a research reagent and synthetic intermediate. It is supplied as a liquid and is employed in organic synthesis workflows, including linker installation and aldehyde-based derivatizations. Recommended handling includes refrigerated storage under inert atmosphere, and colder conditions for solutions to maximize stability.
  • Reactive primary aldehyde suitable for condensation and oxidation reactions.
  • Useful as a linker or protecting-group intermediate in multistep synthesis.
  • Liquid physical form facilitates weighing and solution preparation.
  • Suitable for small-scale screening and preparative chemistry.
  • Stable when stored under nitrogen at 4°C; in solvent, colder storage extends shelf life.
Catalog No. 50-002-83715 Supplier Medchemexpress LLC Supplier No. HYW143230500MG
4-(Benzyloxy)butanal is a benzyloxy-substituted aliphatic aldehyde used as a research reagent and synthetic intermediate. It is supplied as a liquid and is employed in organic synthesis workflows, including linker installation and aldehyde-based derivatizations. Recommended handling includes refrigerated storage under inert atmosphere, and colder conditions for solutions to maximize stability.
  • Reactive primary aldehyde suitable for condensation and oxidation reactions.
  • Useful as a linker or protecting-group intermediate in multistep synthesis.
  • Liquid physical form facilitates weighing and solution preparation.
  • Suitable for small-scale screening and preparative chemistry.
  • Stable when stored under nitrogen at 4°C; in solvent, colder storage extends shelf life.