Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Benzyloxy)phenol, 98%

Catalog No. AC169780010
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC169780010 1 kg Glass bottle
AC169780500 50 g Glass bottle
AC169782500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC169780010 Supplier Thermo Scientific Chemicals Supplier No. 169780010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 103-16-2 | C13H12O2 | 200.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-16-2
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.24
MDL Number MFCD00002333
InChI Key VYQNWZOUAUKGHI-UHFFFAOYSA-N
Synonym 4-benzyloxyphenol, monobenzone, 4-benzyloxy phenol, hydroquinone monobenzyl ether, benoquin, benzoquin, superlite, monobenzyl hydroquinone, leucodinine, monobenzon
PubChem CID 7638
ChEBI CHEBI:34380
IUPAC Name 4-phenylmethoxyphenol
SMILES C1=CC=C(C=C1)COC2=CC=C(C=C2)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 121.0°C to 122.0°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2OC6H4OH
Quantity 1 kg
Beilstein 06,845
Merck Index 15,6334
Solubility Information Solubility in water: slightly soluble. Other solubilities: very soluble in ethanol,benzene,diethyl ether,,soluble in acetone and alkalies,,practically insoluble in petroleum hydrocarbons
Formula Weight 200.24
Percent Purity 98%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material 4-(Benzyloxy)phenol, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause an allergic skin reaction.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-083-3
RTECSNumber SJ7700000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.