Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzyloxybenzaldehyde, TCI America™
SDP

Catalog No. B132525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B132525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B132525G Supplier TCI America Supplier No. B132525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4397-53-9
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD00003387,MFCD01632880
InChI Key ZVTWZSXLLMNMQC-UHFFFAOYSA-N
Synonym 4-benzyloxybenzaldehyde, 4-benzyloxy benzaldehyde, benzaldehyde, 4-phenylmethoxy, p-benzyloxy benzaldehyde, benzaldehyde, p-benzyloxy, 4-benzyloxy-benzaldehyde, 4-phenylmethoxy benzaldehyde, 4-benzyl-oxy benzaldehyde, wang aldehyde resin
PubChem CID 78109
IUPAC Name 4-(benzyloxy)benzaldehyde
SMILES O=CC1=CC=C(OCC2=CC=CC=C2)C=C1

Specifications

Melting Point 74°C
Color White-Yellow
Boiling Point 219°C
Quantity 25 g
Formula Weight 212.25
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzyloxybenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.