Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Benzyloxybutyric acid

Catalog No. 46960225ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-960-225ML 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 46-960-225ML Supplier Sigma Aldrich Supplier No. 46960225ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Benzyloxybutyric acid

Specifications

CAS 10385-30-5
Density 1.097 g/mL (at 25°C (literature))
Boiling Point 135°C (0.3 mmHg)
Molecular Formula C11H14O3
Linear Formula C6H5CH2O(CH2)3 CO2H
Refractive Index n20/D 1.512 (literature)
MDL Number MFCD01321257
Quantity 25 g
Molecular Weight (g/mol) 194.23
Percent Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.