Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Benzyloxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. B47255G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4725-5G 5 g
B47251G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4725-5G Supplier TCI America Supplier No. B47255G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 184000-11-1
Molecular Formula C14H13BO4
Molecular Weight (g/mol) 256.06
MDL Number MFCD02179442
InChI Key UXSDJFGNRVVDFM-UHFFFAOYSA-N
PubChem CID 2773251
IUPAC Name {4-[(benzyloxy)carbonyl]phenyl}boronic acid
SMILES OB(O)C1=CC=C(C=C1)C(=O)OCC1=CC=CC=C1

Specifications

Melting Point 192°C
Color White-Yellow
Quantity 5 g
Synonym 4-benzyloxycarbonylphenylboronic acid, 4-benzyloxycarbonylphenyl boronic acid, 4-benzyloxycarbonyl benzeneboronic acid, 4-benzyloxy carbonyl phenyl boronic acid, 4-benzyloxycarbonyl phenylboronic acid
Formula Weight 256.06
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.