Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzyloxyindole-3-carboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog No. AAL1756703
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1756703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1756703 Supplier Thermo Scientific Chemicals Supplier No. L1756703
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7042-71-9 | C16H13NO2 | 251.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7042-71-9
Molecular Formula C16H13NO2
Molecular Weight (g/mol) 251.29
MDL Number MFCD00152002
InChI Key WURDDLFAUSUSLT-UHFFFAOYSA-N
Synonym 4-benzyloxyindole-3-carboxaldehyde, 4-benzyloxyindole-3-carbaldehyde, 4-benzyloxy-1h-indole-3-carbaldehyde, 4-phenylmethoxy-1h-indole-3-carboxaldehyde, 4-phenylmethoxy indole-3-carbaldehyde, 4-benayloxyindole-3-carboxaldehyde, pubchem7231, acmc-1bcm9, 4-benzyloxy-3-formylindole, 4-benzoxy-1h-indole-3-carbaldehyde
PubChem CID 7015846
IUPAC Name 4-phenylmethoxy-1H-indole-3-carbaldehyde
SMILES O=CC1=CNC2=CC=CC(OCC3=CC=CC=C3)=C12

Specifications

Melting Point 167°C (decomposition)
Quantity 1 g
Beilstein 1537328
Sensitivity Air Sensitive
Formula Weight 251.28
Percent Purity 98%
Chemical Name or Material 4-Benzyloxyindole-3-carboxaldehyde
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.