Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzyloxyphenylacetonitrile 98.0+%, TCI America™
SDP

Catalog No. B47341G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47341G 1 g
B47345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47341G Supplier TCI America Supplier No. B47341G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 838-96-0
Molecular Formula C15H13NO
Molecular Weight (g/mol) 223.28
MDL Number MFCD00016400
InChI Key QKEYZRVDFZDOEP-UHFFFAOYSA-N
Synonym 4-benzyloxyphenylacetonitrile, 2-4-benzyloxy phenyl acetonitrile, 4-benzyloxybenzyl cyanide, 4-benzyloxy phenyl acetonitrile, 2-4-phenylmethoxy phenyl ethanenitrile, acmc-20amoi, pubchem15926, 4-benzyloxybenzeneacetonitrile, p-benzyloxyphenyl acetonitrile
PubChem CID 522804
IUPAC Name 2-[4-(benzyloxy)phenyl]acetonitrile
SMILES N#CCC1=CC=C(OCC2=CC=CC=C2)C=C1

Specifications

Melting Point 68°C
Boiling Point 173°C
Quantity 1 g
Formula Weight 223.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzyloxyphenylacetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.