Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzyloxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. B21451G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21451G 1 g
B2145-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21451G Supplier TCI America Supplier No. B21451G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 146631-00-7
Molecular Formula C13H13BO3
Molecular Weight (g/mol) 228.054
MDL Number MFCD01075705
InChI Key DMJHEIDWSIAXCS-UHFFFAOYSA-N
PubChem CID 2734314
IUPAC Name (4-phenylmethoxyphenyl)boronic acid
SMILES B(C1=CC=C(C=C1)OCC2=CC=CC=C2)(O)O

Specifications

Quantity 1 g
Synonym 4-benzyloxyphenylboronic acid, 4-benzyloxybenzeneboronic acid, 4-benzyloxy phenyl boronic acid, 4-benzyloxy phenylboronic acid, 4-benzyloxyphenyl boronic acid, 4-benzyl-oxy phenyl boranediol, boronic acid, 4-phenylmethoxy phenyl, 4-phenylmethoxyphenyl boronic acid, pubchem9561
Formula Weight 228.05
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzyloxyphenylboronic Acid (contains varying amounts of Anhydride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.