Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Benzyloxypropiophenone, 98%, Thermo Scientific™

Catalog No. AAA1832730
Change view
Click to view available options
Quantity:
10 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1832730 250 g
AAA1832709 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1832730 Supplier Thermo Scientific Chemicals Supplier No. A1832730

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4495-66-3
Molecular Formula C16H16O2
Molecular Weight (g/mol) 240.30
MDL Number MFCD00009311
InChI Key IKFGSOJYHVTNDV-UHFFFAOYSA-N
Synonym 4-benzyloxypropiophenone, 1-4-benzyloxy phenyl propan-1-one, p-benzyloxypropiophenone, 4-benzyloxy-propiophenone, 4'-benzyloxypropiophenone, 1-4-benzyloxy phenyl-1-propanone, 1-propanone, 1-4-phenylmethoxy phenyl, 1-4-phenylmethoxy phenyl propan-1-one, 4-benzyloxy propiophenone
PubChem CID 78246
SMILES CCC(=O)C1=CC=C(OCC2=CC=CC=C2)C=C1

Specifications

Melting Point 98°C to 102°C
Quantity 250 g
Beilstein 2271608
IUPAC Name 1-(4-phenylmethoxyphenyl)propan-1-one
Formula Weight 240.3
Percent Purity 98%
Chemical Name or Material 4'-Benzyloxypropiophenone
Hazard Category H500
EINECSNumber 224-788-2
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.