Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Benzyloxypropiophenone, 99%, Thermo Scientific™

Catalog No. AC165582500
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC165582500 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC165582500 Supplier Thermo Scientific Chemicals Supplier No. 165582500

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4495-66-3
Molecular Formula C16H16O2
Molecular Weight (g/mol) 240.30
MDL Number MFCD00009311
InChI Key IKFGSOJYHVTNDV-UHFFFAOYSA-N
Synonym 4-benzyloxypropiophenone, 1-4-benzyloxy phenyl propan-1-one, p-benzyloxypropiophenone, 4-benzyloxy-propiophenone, 4'-benzyloxypropiophenone, 1-4-benzyloxy phenyl-1-propanone, 1-propanone, 1-4-phenylmethoxy phenyl, 1-4-phenylmethoxy phenyl propan-1-one, 4-benzyloxy propiophenone
PubChem CID 78246
IUPAC Name 1-[4-(benzyloxy)phenyl]propan-1-one
SMILES CCC(=O)C1=CC=C(OCC2=CC=CC=C2)C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 99°C to 102°C
Color White to Beige
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula C6H5CH2OC6H4COC2H5
Quantity 250 g
Formula Weight 240.3
Percent Purity ≥98.5%
Physical Form Powder
Chemical Name or Material 4'-Benzyloxypropiophenone, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.