Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzylpiperidine 99.0+%, TCI America™
SDP

Catalog No. B0387250ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0387250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B0387250ML Supplier TCI America Supplier No. B0387250ML

missing translation for 'validMainframeMsg'

Specifications

CAS 31252-42-3
Color Yellow
Boiling Point 279°C
Molecular Formula C12H17N
Quantity 250 mL
MDL Number MFCD00006006
Synonym piperidine, 4-phenylmethyl, 4-phenylmethyl piperidine, 4-benzyl-piperidine, piperidine, 4-benzyl, phenyl 4-piperidyl methane, alpha-4-piperidyl toluene, unii-jdf1t4667s, 4-benzyl piperidine, 4-benzylpiperdine, 4-benzylpiperidin
InChI Key ABGXADJDTPFFSZ-UHFFFAOYSA-N
SMILES C(C1CCNCC1)C1=CC=CC=C1
IUPAC Name 4-benzylpiperidine
Molecular Weight (g/mol) 175.28
PubChem CID 31738
Formula Weight 175.28
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Benzylpiperidine
Show More Show Less
RTECSNumber TM4728000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.