Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzylresorcinol 97.0+%, TCI America™
SDP

Catalog No. B04385G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B04385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B04385G Supplier TCI America Supplier No. B04385G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2284-30-2
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.237
MDL Number MFCD00059620
InChI Key QVFIWTNWKHFVEH-UHFFFAOYSA-N
PubChem CID 75294
IUPAC Name 4-benzylbenzene-1,3-diol
SMILES C1=CC=C(C=C1)CC2=C(C=C(C=C2)O)O

Specifications

Melting Point 79°C
Boiling Point 215°C
Quantity 5 g
Formula Weight 200.24
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzylresorcinol
RTECSNumber VH0175000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.