Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Biphenylacetonitrile, 97%, Thermo Scientific™

Catalog No. AC443790250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC443790250 25 g
AC443790010 1 g
AC443790050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC443790250 Supplier Thermo Scientific Chemicals Supplier No. 443790250

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 31603-77-7
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.25
MDL Number MFCD00016403
InChI Key HSZCNGTZJWZAMF-UHFFFAOYSA-N
Synonym 4-biphenylacetonitrile, 2-1,1'-biphenyl-4-yl acetonitrile, 2-4-phenylphenyl acetonitrile, 1,1'-biphenyl-4-acetonitrile, 4-cyanomethylbiphenyl, p-biphenylacetonitrile, p-biphenylylacetonitrile, 4-biphenyl-acetonitrile, 4-biphenylylacetonitrile, p-phenyl benzyl cyanide
PubChem CID 35856
IUPAC Name 2-(4-phenylphenyl)acetonitrile
SMILES N#CCC1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 95.0°C to 97.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 25 g
Formula Weight 193.25
Percent Purity 97%
Chemical Name or Material 4-Biphenylacetonitrile
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.