Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(BOC-amino)-1-butanol, 98%

Catalog No. AC434210050
Change view
Click to view available options
Quantity:
1 mL
5 mL
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC434210050 5 mL Glass Bottle
AC434210010 1 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC434210050 Supplier Thermo Scientific Chemicals Supplier No. 434210050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 75178-87-9 | C9H19NO3 | 189.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75178-87-9
Molecular Formula C9H19NO3
Molecular Weight (g/mol) 189.26
MDL Number MFCD01861334
InChI Key LIYMTLVBAVHPBU-UHFFFAOYSA-N
Synonym tert-butyl 4-hydroxybutyl carbamate, tert-butyl n-4-hydroxybutyl carbamate, 4-boc-amino-1-butanol, 4-tert-butoxycarbonylamino-1-butanol, carbamic acid, 4-hydroxybutyl-, 1,1-dimethylethyl ester, tert-butyl 4-hydroxybutylcarbamate, n-boc-4-aminobutanol, butylhydroxybutylcarbamate, acmc-1bfe1, 4-t-boc-amino-1-butanol
PubChem CID 545141
IUPAC Name tert-butyl N-(4-hydroxybutyl)carbamate
SMILES CC(C)(C)OC(=O)NCCCCO

Specifications

Melting Point 37°C to 40°C
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Density 1.0200g/mL
Quantity 5 mL
Formula Weight 189.25
Percent Purity 98%
Specific Gravity 1.02
Physical Form Low Melting Solid or Liquid
Chemical Name or Material 4-(Boc-Amino)-1-butanol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.