Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Boc-amino)-4-carbamoylbutyric acid, 98%

Catalog No. AAH62305MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62305MD 250 mg
AAH6230503 1 g
AAH6230506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH62305MD Supplier Thermo Scientific Chemicals Supplier No. H62305MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 18800-74-3 | C10H17N2O5 | 245.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18800-74-3
Molecular Formula C10H17N2O5
Molecular Weight (g/mol) 245.26
MDL Number MFCD00070081
InChI Key GMZBDBBBJXGPIH-LURJTMIESA-M
Synonym boc-glu-nh2, n-alpha-tert-boc-l-isoglutamine, 4s-4-n-tert-butoxycarbonylamino-4-carbamoyl butanoic acid, 4s-4-tert-butoxycarbonyl amino-4-carbamoylbutanoic acid, boc-glu-nh, ambotzbaa1095, na-t-boc-l-isoglutamine, boc-isogln-oh;boc-l-isoglutamine, n-alpha-t-butyloxycarbonyl-l-glutamic amide, 4-boc-amino-4-carbamoylbutyric acid
PubChem CID 2734757
IUPAC Name (4S)-5-amino-4-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](CCC([O-])=O)C(N)=O

Specifications

Color White
Quantity 250 mg
Formula Weight 246.26
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-(Boc-amino)-4-carbamoylbutyric acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.