Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Boc-amino)phenol, 97%

Catalog No. p-7037074
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5564506 5 g
AAH5564514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5564506 Supplier Thermo Scientific Chemicals Supplier No. H5564506
Only null left
Add to Cart

CAS: 54840-15-2 | C11H15NO3 | 209.245 g/mol

4-(Boc-amino)phenol, is used to protect amine in the solid phase synthesis of peptides, used to produce [4-(tert-butyl-dimethyl-silanyloxy)-phenyl]-carbamic acid tert-butyl ester at ambient temperature. This reaction will need reagent imidazole and solvent dimethylformamide, CH2Cl2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Boc-amino)phenol, is used to protect amine in the solid phase synthesis of peptides, used to produce [4-(tert-butyl-dimethyl-silanyloxy)-phenyl]-carbamic acid tert-butyl ester at ambient temperature. This reaction will need reagent imidazole and solvent dimethylformamide, CH2Cl2.

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54840-15-2
Molecular Formula C11H15NO3
Molecular Weight (g/mol) 209.245
MDL Number MFCD00226573
InChI Key YRQMBQUMJFVZLF-UHFFFAOYSA-N
Synonym 4-n-boc-aminophenol, tert-butyl 4-hydroxyphenyl carbamate, tert-butyl n-4-hydroxyphenyl carbamate, tert-butyl 4-hydroxyphenylcarbamate, 4-boc-amino phenol, n-boc-4-hydroxyaniline, 1,1-dimethylethyl 4-hydroxyphenyl carbamate, 4-tert-butoxycarbonylamino phenol, 4-hydroxy-phenyl-carbamic acid tert-butyl ester, carbamic acid, 4-hydroxyphenyl-, 1,1-dimethylethyl ester
PubChem CID 2756771
IUPAC Name tert-butyl N-(4-hydroxyphenyl)carbamate
SMILES CC(C)(C)OC(=O)NC1=CC=C(C=C1)O

Specifications

Melting Point 143°C to 147°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 209.25
Percent Purity 97%
Chemical Name or Material 4-(Boc-amino)phenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.