Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Boc-amino)pyridine, 95%

Catalog No. AAH3082014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3082014 25 g
AAH3082006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3082014 Supplier Thermo Scientific Chemicals Supplier No. H3082014
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 98400-69-2 | C10H14N2O2 | 194.234 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98400-69-2
Molecular Formula C10H14N2O2
Molecular Weight (g/mol) 194.234
MDL Number MFCD02179232
InChI Key DRZYCRFOGWMEES-UHFFFAOYSA-N
Synonym 4-boc-amino pyridine, 4-tert-butoxycarbonylamino pyridine, tert-butyl pyridin-4-ylcarbamate, tert-butyl n-pyridin-4-yl carbamate, tert-butyl n-4-pyridyl carbamate, carbamic acid, 4-pyridinyl-, 1,1-dimethylethyl ester, pyridin-4-yl-carbamic acid tert-butyl ester, pubchem20060, acmc-209sa9, ksc495m3f
PubChem CID 9990210
IUPAC Name tert-butyl N-pyridin-4-ylcarbamate
SMILES CC(C)(C)OC(=O)NC1=CC=NC=C1

Specifications

Melting Point 147°C to 150°C
Quantity 25 g
Formula Weight 194.23
Percent Purity 95%
Chemical Name or Material 4-(Boc-amino)pyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.