Learn More
Medchemexpress LLC (4-Boc-aminophenyl)boronic acid | 380430-49-9 | 237.06 | 10 G
SDP
Catalog No. 5000447536
Encompass_Preferred
(4-Boc-aminophenyl)boronic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only.
  • Biochemical reagent
  • Suitable for life science related research
  • Solid appearance
  • White to off-white color
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-47536 Supplier Medchemexpress LLC Supplier No. HYW00092310G
(4-Boc-aminophenyl)boronic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only.
  • Biochemical reagent
  • Suitable for life science related research
  • Solid appearance
  • White to off-white color
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month