Learn More
Medchemexpress LLC 4-borono-L-phenylalanine | 76410-58-7 | MFCD01075172 | 99.8% | 209.01 g/mol | C9H12BNO4 | 1 G
SDP
Catalog No. 5000208542
Encompass_Preferred
L-p-boronophenylalanine (4-borono-L-phenylalanine) is a boron-containing amino acid used in research to selectively deliver boron to tumor cells for boron neutron capture therapy (BNCT). It is a substrate for L-type amino acid transporters and accumulates in certain cancers such as glioblastoma.
  • Substrate for L-type amino acid transporters (LAT1 and LAT2).
  • Used in research on boron neutron capture therapy (BNCT).
  • High purity suitable for analytical and research applications.
  • Solid form with defined storage conditions to maintain stability.
Catalog No. 50-002-08542 Supplier Medchemexpress LLC Supplier No. HYW0878301G
L-p-boronophenylalanine (4-borono-L-phenylalanine) is a boron-containing amino acid used in research to selectively deliver boron to tumor cells for boron neutron capture therapy (BNCT). It is a substrate for L-type amino acid transporters and accumulates in certain cancers such as glioblastoma.
  • Substrate for L-type amino acid transporters (LAT1 and LAT2).
  • Used in research on boron neutron capture therapy (BNCT).
  • High purity suitable for analytical and research applications.
  • Solid form with defined storage conditions to maintain stability.