Learn More
Medchemexpress LLC 4-Br-Bnlm | 1654775-71-9 | 99.5% | 50 MG
SDP
Catalog No. 5000231078
Encompass_Preferred
4-Br-Bnlm is a small-molecule inhibitor of glucose-regulated protein 94 (Grp94) supplied for research use. It has been used to promote clearance of toxic myocilin forms and to reduce myocilin-associated cellular toxicity. For research use only; not for human use.
  • Selective Grp94 inhibitor
  • Promotes clearance of toxic myocilin forms
  • High reported chemical purity
  • Multiple pack sizes for experimental flexibility
  • Stable when stored under recommended conditions
Catalog No. 50-002-31078 Supplier Medchemexpress LLC Supplier No. HY12184550MG
4-Br-Bnlm is a small-molecule inhibitor of glucose-regulated protein 94 (Grp94) supplied for research use. It has been used to promote clearance of toxic myocilin forms and to reduce myocilin-associated cellular toxicity. For research use only; not for human use.
  • Selective Grp94 inhibitor
  • Promotes clearance of toxic myocilin forms
  • High reported chemical purity
  • Multiple pack sizes for experimental flexibility
  • Stable when stored under recommended conditions