Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-1,3,5-trimethylpyrazole 98.0+%, TCI America™
SDP

Catalog No. B47445G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47445G 5 g
B47441G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47445G Supplier TCI America Supplier No. B47445G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15801-69-1
Molecular Formula C6H9BrN2
Molecular Weight (g/mol) 189.056
MDL Number MFCD00052532
InChI Key UNTQXOJGXGRHMG-UHFFFAOYSA-N
Synonym 4-bromo-1,3,5-trimethyl-1h-pyrazole, 4-bromo-2,3,5-trimethylpyrazole, 4-bromo-1,3,5-trimethyl-2h-pyrazole, pubchem20507, acmc-209dhc, buttpark 34\09-42, 4-bromo-1,3,5-trimethylpyrazole, 4-bromo-1,3,5-trimethyl-pyrazole, 4-bromo-1,3,5-trimethyl pyrazole
PubChem CID 519181
IUPAC Name 4-bromo-1,3,5-trimethylpyrazole
SMILES CC1=C(C(=NN1C)C)Br

Specifications

Melting Point 36°C
Color White-Yellow
Boiling Point 97°C
Quantity 5 g
Formula Weight 189.06
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Bromo-1,3,5-trimethylpyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.