Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-1,8-naphthalic Anhydride 95.0+%, TCI America™
SDP

Catalog No. B085825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B085825G 25 g
B08585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B085825G Supplier TCI America Supplier No. B085825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 81-86-7
Molecular Formula C12H5BrO3
Molecular Weight (g/mol) 277.073
MDL Number MFCD00006927
InChI Key DTUOTSLAFJCQHN-UHFFFAOYSA-N
Synonym 4-Bromo-1,8-naphthalenedicarboxylic Anhydride
PubChem CID 66493
SMILES C1=CC2=C(C=CC3=C2C(=C1)C(=O)OC3=O)Br

Specifications

Melting Point 221°C
Color White-Yellow
Quantity 25 g
Formula Weight 277.07
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-1,8-naphthalic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.