Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Bromo-1-butene

Catalog No. 16785110G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-785-110G 10 g
16-785-11G 1 g
16-785-150G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 16-785-110G Supplier Sigma Aldrich Supplier No. 16785110G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Stereoselective epoxidation of 4-bromo-1-butene was carried out with three alkene-utilizing bacteria: mycobacterium li, mycobacterium e3 and nocardia ip1
  • 4-bromo-1-butene was used in the synthesis of 4-(3-butenyloxy)benzoic acid
  • It was also used in the total synthesis of largazole
  • It was used in double alkylation of lactams, followed by grubbs ring-closing metathesis, to yield gymnodimine (biotoxin)
  • It was used in preparation of dienehydrazides

Specifications

CAS 5162-44-7
Density 1.33 g/mL (at 25°C (literature))
Boiling Point 98°C to 100°C (lit.)
Molecular Formula C4H7Br
Refractive Index n20/D 1.462 (literature)
Linear Formula BrCH2CH2CH=CH2
MDL Number MFCD00000258
Quantity 10 g
Molecular Weight (g/mol) 135
Percent Purity 97%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.