Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-1-butene, 98+%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
50mL
Packaging:
Glass bottle
5162-44-7
C4H7Br
135
MFCD00000258
DMAYBPBPEUFIHJ-UHFFFAOYSA-N
4-bromo-1-butene,3-butenyl bromide,1-butene, 4-bromo,4-bromo-1-buten,4-bromo-but-1-ene,homoallyl bromide,1-bromo-3-butene,4-bromobutene,4-bromo-1 butene,sjphabijuh@
21241
4-bromobut-1-ene
C=CCCBr
5162-44-7
C4H7Br
135
MFCD00000258
DMAYBPBPEUFIHJ-UHFFFAOYSA-N
4-bromo-1-butene,3-butenyl bromide,1-butene, 4-bromo,4-bromo-1-buten,4-bromo-but-1-ene,homoallyl bromide,1-bromo-3-butene,4-bromobutene,4-bromo-1 butene,sjphabijuh@
21241
4-bromobut-1-ene
C=CCCBr
Specific Gravity 1.33
Packaging Glass bottle
Refractive Index 1.4595 to 1.4635
Quantity 25mL
Flash Point 1°C
Boiling Point 98.0°C to 100.0°C
Density 1.3300g/mL
Infrared Spectrum Authentic
Color Brown to Colorless
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Linear Formula BrCH2CH2CH=CH2
Beilstein 01, I, 84
Formula Weight 135
Physical Form Liquid
Percent Purity 98+%
Chemical Name or Material 4-Bromo-1-butene, 98+%
Show More Show Less

GHS H Statement
Causes serious eye irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
May cause an allergic skin reaction.
Highly flammable liquid and va

GHS P Statement
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear protective gloves/pr

GHS Signal Word: Danger

EINECSNumber : 225-937-4

TSCA : TSCA