Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-1-butene, 98+%, Thermo Scientific™

Catalog No. AC173550100
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC173550100 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC173550100 Supplier Thermo Scientific Chemicals Supplier No. 173550100

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5162-44-7
Molecular Formula C4H7Br
Molecular Weight (g/mol) 135.004
InChI Key DMAYBPBPEUFIHJ-UHFFFAOYSA-N
Synonym 4-bromo-1-butene, 3-butenyl bromide, 1-butene, 4-bromo, 4-bromo-1-buten, 4-bromo-but-1-ene, homoallyl bromide, 1-bromo-3-butene, 4-bromobutene, 4-bromo-1 butene, sjphabijuh@
PubChem CID 21241
IUPAC Name 4-bromobut-1-ene
SMILES C=CCCBr

Specifications

Density 1.3300g/mL
Boiling Point 98.0°C to 100.0°C
Flash Point 1°C
Linear Formula BrCH2CH2CH=CH2
Refractive Index 1.4595 to 1.4635
Quantity 10 mL
Formula Weight 135
Chemical Name or Material 4-Bromo-1-butene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.