Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-1-methyl-1H-indole, 95%, Thermo Scientific™

Catalog No. AAH66535MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66535MD 250 mg
AAH6653503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66535MD Supplier Thermo Scientific Chemicals Supplier No. H66535MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 590417-55-3
Molecular Formula C9H8BrN
Molecular Weight (g/mol) 210.07
MDL Number MFCD08435875
InChI Key JZOSXTYDJPHXQD-UHFFFAOYSA-N
Synonym 4-bromo-1-methyl-1h-indole, 4-bromo-1-methyl-indole, acmc-209m9m, 1h-indole,4-bromo-1-methyl, 4-bromo-1-methylindole, 1h-indole, 4-bromo-1-methyl
PubChem CID 22558937
IUPAC Name 4-bromo-1-methyl-1H-indole
SMILES CN1C=CC2=C(Br)C=CC=C12

Specifications

Color Colorless to Yellow
Density 1.456 g/mL
Flash Point 110°C (230°F)
Refractive Index 1.636
Quantity 250 mg
Formula Weight 210.06
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 4-Bromo-1-methyl-1H-indole
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.