Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Bromo-1-methyl-1H-pyrazole

Catalog No. 69645525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-645-525G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 69-645-525G Supplier Sigma Aldrich Supplier No. 69645525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Bromo-1-methyl-1H-pyrazole

Specifications

CAS 15803-02-8
Density 1.558 g/mL (at 25°C)
Boiling Point 185°C to 188°C (760 mmHg)
Molecular Formula C4H5BrN2
Refractive Index n20/D 1.531
Linear Formula C4H5BrN2
MDL Number MFCD02179565
Quantity 250 mg
Synonym 1-Methyl-4-bromopyrazole
Molecular Weight (g/mol) 161
Percent Purity 97%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.