Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-1-naphthaleneboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. B44631G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B44631G Supplier TCI America Supplier No. B44631G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 145965-14-6
Molecular Formula C10H8BBrO2
Molecular Weight (g/mol) 250.89
MDL Number MFCD08701764
InChI Key DKKOMUVRCPKTFX-UHFFFAOYSA-N
Synonym 4-bromonaphthalen-1-yl boronic acid, 4-bromonaphthalene-1-boronic acid, 4-bromo-1-naphthaleneboronic acid, boronic acid, 4-bromo-1-naphthalenyl, 4-bromo-1-naphthalenyl-boronic acid, acmc-209cw4, ksc524o6f, 4-bromo-1-naphthylboronic acid, 1-bromonaphthalene-4-boronic acid, 1-bromo-4-dihydroxyboryl naphthalene
PubChem CID 23005424
IUPAC Name (4-bromonaphthalen-1-yl)boronic acid
SMILES OB(O)C1=C2C=CC=CC2=C(Br)C=C1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 250.89
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-1-naphthaleneboronic Acid (contains varying amounts of Anhydride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.