Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Bromo-1H-indazole | 186407-74-9 | 99.98% | C7H5BrN2 | 10 G
SDP

Catalog No. 5000434249
Encompass_Preferred

4-Bromo-1H-indazole is a biochemical reagent. It can be utilized as a biological material or organic compound for diverse applications in life science related research.

  • High purity: 99.98%
  • Appearance: Off-white to light brown solid
  • Molecular weight: 197.03
  • Soluble in DMSO (100 mg/mL)
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • Solution storage: -80°C for 6 months, -20°C for 1 month
  • Compatible with various in vivo dissolution protocols

Catalog No. 50-004-34249 Supplier Medchemexpress LLC Supplier No. HY6908510G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Bromo-1H-indazole is a biochemical reagent. It can be utilized as a biological material or organic compound for diverse applications in life science related research.

  • High purity: 99.98%
  • Appearance: Off-white to light brown solid
  • Molecular weight: 197.03
  • Soluble in DMSO (100 mg/mL)
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • Solution storage: -80°C for 6 months, -20°C for 1 month
  • Compatible with various in vivo dissolution protocols

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.