Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-1H-pyrazole-5-carbaldehyde, 95+%, Thermo Scientific™

Catalog No. RF03950DE
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RF03950DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RF03950DE Supplier Thermo Scientific Chemicals Supplier No. RF03950DE
Add to Cart

Chemical Identifiers

CAS 287917-97-9
Molecular Formula C4H3BrN2O
Molecular Weight (g/mol) 174.985
InChI Key UWGFONONBAIDAF-UHFFFAOYSA-N
Synonym 4-bromo-1h-pyrazole-3-carbaldehyde, 4-bromo-1h-pyrazole-5-carboxaldehyde, 4-bromo-2h-pyrazole-3-carbaldehyde, 4-bromo-1h-pyrazole-3-carboxaldehyde, 1h-pyrazole-3-carboxaldehyde,4-bromo, 1h-pyrazole-3-carboxaldehyde, 4-bromo, 4-bromopyrazole-5-carbaldehyde, 4-bromo-5-formyl-1h-pyrazole, 4-bromo-pyrazole-3-carboxaldehyde, 4-bromo-2h-pyrazole-3-carboxaldehyde
PubChem CID 2782154
IUPAC Name 4-bromo-1H-pyrazole-5-carbaldehyde
SMILES C1=NNC(=C1Br)C=O

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 174.98
Percent Purity 95%
Chemical Name or Material 4-Bromo-1H-pyrazole-5-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.