Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-(1H-pyrazol-3-yl)phenol 98.0+%, TCI America™
SDP

Catalog No. B47431G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47431G 1 g
B47435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47431G Supplier TCI America Supplier No. B47431G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 99067-15-9
Molecular Formula C9H7BrN2O
Molecular Weight (g/mol) 239.072
MDL Number MFCD03094026
InChI Key WAHULEHSSFNFQX-UHFFFAOYSA-N
Synonym 4-bromo-2-1h-pyrazol-3-yl phenol, 4-bromo-6-1,2-dihydropyrazol-3-ylidene cyclohexa-2,4-dien-1-one, 2-1h-pyrazole-3-yl-4-bromophenol, 4-bromo-2-2h-pyrazol-3-yl phenol
PubChem CID 6744501
IUPAC Name 4-bromo-6-(1,2-dihydropyrazol-3-ylidene)cyclohexa-2,4-dien-1-one
SMILES C1=CC(=O)C(=C2C=CNN2)C=C1Br

Specifications

Melting Point 160°C
Color Purple-White
Quantity 1 g
Formula Weight 239.07
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-(1H-pyrazol-3-yl)phenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.