Learn More
Medchemexpress LLC 4-bromo-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione | 2093536-12-8 | 99.6% | 337.13 g/mol | C13H9BrN2O4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW039116100MG
Encompass_Preferred
Thalidomide-4-Br is a brominated thalidomide analog used as a research reagent and cereblon/E3 ligase ligand. It is employed in PROTAC development and in studies of inflammation, tumor biology, and neurology. The compound is supplied as a solid with high reported purity and should be stored cold according to handling recommendations.
  • Useful as an E3 ligase and cereblon ligand in PROTAC design and degradation studies.
  • Applicable to research in inflammation, tumor biology, and neurology.
  • Provided as an off-white to gray solid in multiple pack sizes suitable for screening and synthesis.
  • High reported purity (99.6%).
  • Stable when stored cold; follow recommended powder and solvent storage conditions.
  • Available in solution format for screening, for example 10 mM in DMSO.
Catalog No. 50-003-01693