Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2,3,5,6-tetrafluorobenzoic acid, 97+%

Catalog No. AAH3298203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3298203 1 g
AAH3298206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3298203 Supplier Thermo Scientific Chemicals Supplier No. H3298203
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 4707-24-8 | C7HBrF4O2 | 272.981 g/mol

4-Bromo-2,3,5,6-tetrafluorobenzoic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2,3,5,6-tetrafluorobenzoic acid is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4707-24-8
Molecular Formula C7HBrF4O2
Molecular Weight (g/mol) 272.981
MDL Number MFCD00075244
InChI Key MTZLICDXLGQWJV-UHFFFAOYSA-N
Synonym 4-bromo-2,3,5,6-tetrafluoro-benzoic acid, acmc-1arsf, 4-bromotetrafluorobenzoic acid, 4-bromotetrafluorobenzene carboxylic acid, benzoicacid, 4-bromo-2,3,5,6-tetrafluoro, benzoic acid, 4-bromo-2,3,5,6-tetrafluoro, 4-bromo-2,3,5,6-tetrafluorobenzoic acid, 4-bromanyl-2,3,5,6-tetrakis fluoranyl benzoic acid
PubChem CID 298173
IUPAC Name 4-bromo-2,3,5,6-tetrafluorobenzoic acid
SMILES C1(=C(C(=C(C(=C1F)F)Br)F)F)C(=O)O

Specifications

Melting Point 142°C to 145°C
Odor Odorless
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 272.98
Percent Purity ≥97%
Chemical Name or Material 4-Bromo-2,3,5,6-tetrafluorobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.