Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2,3,5,6-tetrafluorobenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog No. AAL2015306
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2015306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL2015306 Supplier Thermo Scientific Chemicals Supplier No. L2015306
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 122033-54-9 | Molecular Formula: C7BrClF4O | Molecular Weight: 291.424 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122033-54-9
Molecular Formula C7BrClF4O
Molecular Weight (g/mol) 291.424
MDL Number MFCD03001173
InChI Key XVYIQLOULDERAO-UHFFFAOYSA-N
Synonym 4-bromo-2,3,5,6-tetrafluorobenzoylchloride, benzoyl chloride,4-bromo-2,3,5,6-tetrafluoro, acmc-20amnz, 4-bromanyl-2,3,5,6-tetrakis fluoranyl benzoyl chloride
PubChem CID 2783245
IUPAC Name 4-bromo-2,3,5,6-tetrafluorobenzoyl chloride
SMILES C1(=C(C(=C(C(=C1F)F)Br)F)F)C(=O)Cl

Specifications

Boiling Point 95°C to 100°C
Refractive Index 1.517
Quantity 5 g
UN Number UN3265
Beilstein 7295811
Sensitivity Moisture sensitive
Formula Weight 291.43
Percent Purity 98%
Chemical Name or Material 4-Bromo-2,3,5,6-tetrafluorobenzoyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.