Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2,6-difluoroaniline 98.0+%, TCI America™
SDP

Catalog No. B32205G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32205G 5 g
B32201G 1 g
B322025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B32205G Supplier TCI America Supplier No. B32205G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67567-26-4
Molecular Formula C6H4BrF2N
Molecular Weight (g/mol) 208.006
MDL Number MFCD00013389
InChI Key BFQSQUAVMNHOEF-UHFFFAOYSA-N
Synonym 2,6-difluoro-4-bromoaniline, 4-bromo-2,6-difluorobenzenamine, benzenamine, 4-bromo-2,6-difluoro, 4-bromo-2,6-difluoro-phenylamine, 4-bromo-2,6-difluoro-aniline, 4-bromo-2,6-difluorophenylamine, buttpark 22\07-66, 4-bromo-2,6-difluorophenyl amine, pubchem2929, acmc-1begp
PubChem CID 610191
IUPAC Name 4-bromo-2,6-difluoroaniline
SMILES C1=C(C=C(C(=C1F)N)F)Br

Specifications

Melting Point 64°C
Quantity 5 g
Formula Weight 208.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2,6-difluoroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.