Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2,6-difluorobenzaldehyde 98.0+%, TCI America™
SDP

Catalog No. B3446100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3446100MG 100 mg
B34461G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B3446100MG Supplier TCI America Supplier No. B3446100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 537013-51-7
Molecular Formula C7H3BrF2O
Molecular Weight (g/mol) 221.00
MDL Number MFCD03094459
InChI Key CZGVAISJIQNQEJ-UHFFFAOYSA-N
Synonym 4-bromo-2,6-difluorobenzylaldehyde, 4-bromo-2,6-difluoro-benzaldehyde, benzaldehyde, 4-bromo-2,6-difluoro, 2,6-difluoro-4-bromobenzaldehyde, 2,6-difluoro-4-bromo benzaldehyde, pubchem4165, acmc-209l9s, ksc911q5f, 4-bromo 2,6-difluorobenzaldehyde, 2,6-difluoro-4-bromo-benzaldehyde
PubChem CID 2773290
IUPAC Name 4-bromo-2,6-difluorobenzaldehyde
SMILES FC1=CC(Br)=CC(F)=C1C=O

Specifications

Melting Point 77°C
Quantity 100 mg
Formula Weight 221.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2,6-difluorobenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.