Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-chlorotoluene 97.0+%, TCI America™
SDP

Catalog No. B20685G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20685G 5 g
B206825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20685G Supplier TCI America Supplier No. B20685G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 89794-02-5
Molecular Formula C7H6BrCl
Molecular Weight (g/mol) 205.479
MDL Number MFCD00060651
InChI Key LIFMTDJMLRECMX-UHFFFAOYSA-N
Synonym 4-bromo-2-chlorotoluene, 2-chloro-4-bromotoluene, 4-bromo-2-chloro toluene, 4-bromo-2-chloro-1-methyl-benzene, benzene, 4-bromo-2-chloro-1-methyl, 4-bromo-chlorotoluene, pubchem3603, 4-bromo-2chlorotoluene, 2-chloro 4-bromo toluene, acmc-209r2x
PubChem CID 2735554
IUPAC Name 4-bromo-2-chloro-1-methylbenzene
SMILES CC1=C(C=C(C=C1)Br)Cl

Specifications

Quantity 5 g
Physical Form Clear Liquid at 20°C
Boiling Point 110°C
Formula Weight 205.48
Percent Purity ≥97.0% (GC)
Chemical Name or Material 4-Bromo-2-chlorotoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.